accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OHONOOCl

CAS RN 84942-40-5

IUPAC Name: 1-(5-chloro-2-hydroxy-3-nitrophenyl)ethan-1-one
Synonyms: 1-(5-chloro-2-hydroxy-3-nitrophenyl)ethanone
Molecular Weight (g/mol): 215.59
Molecular Formula: C8H6ClNO4
InChi Key: IUNBIQBAYUBIFD-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(Cl)=CC(=C1O)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results