CAS RN 85-01-8
Phenanthrene, 98%
CAS: 85-01-8 Molecular Formula: C14H10 Molecular Weight (g/mol): 178.23 MDL Number: MFCD00001168 InChI Key: YNPNZTXNASCQKK-UHFFFAOYSA-N Synonym: phenanthren,phenanthrin,phenanthracene,ravatite,phenantrin,phenanthren german,phenanthrene, pure,unii-448j8e5bst,ccris 1233,chembl46730 PubChem CID: 995 ChEBI: CHEBI:28851 IUPAC Name: phenanthrene SMILES: C1=CC=C2C(C=CC3=CC=CC=C23)=C1
Phenanthrene, 97%
CAS: 85-01-8 Molecular Formula: C14H10 Molecular Weight (g/mol): 178.23 MDL Number: MFCD00001168 InChI Key: YNPNZTXNASCQKK-UHFFFAOYSA-N Synonym: phenanthren,phenanthrin,phenanthracene,ravatite,phenantrin,phenanthren german,phenanthrene, pure,unii-448j8e5bst,ccris 1233,chembl46730 PubChem CID: 995 ChEBI: CHEBI:28851 IUPAC Name: phenanthrene SMILES: C1=CC=C2C(C=CC3=CC=CC=C23)=C1
Phenanthrene in Acetonitrile 100μg/mL, Fisher Chemical™
Phenanthrene, tech. 90%
CAS: 85-01-8 Molecular Formula: C14H10 Molecular Weight (g/mol): 178.23 MDL Number: MFCD00001168 InChI Key: YNPNZTXNASCQKK-UHFFFAOYSA-N Synonym: phenanthren,phenanthrin,phenanthracene,ravatite,phenantrin,phenanthren german,phenanthrene, pure,unii-448j8e5bst,ccris 1233,chembl46730 PubChem CID: 995 ChEBI: CHEBI:28851 IUPAC Name: phenanthrene SMILES: C1=CC=C2C(C=CC3=CC=CC=C23)=C1
Phenanthrene, TRC
CAS: 85-01-8 Molecular Formula: C14 H10 Molecular Weight (g/mol): 178.23 Synonym: Phenanthrene,NSC 26256,Ravatite,[3]Helicene IUPAC Name: phenanthrene SMILES: c1ccc2c(c1)ccc3ccccc23