accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 850567-21-4

BOHNCH3HOOH

CAS RN 850567-21-4

IUPAC Name: [3-(ethylcarbamoyl)phenyl]boronic acid
Synonyms: 3-(ethylcarbamoyl)phenylboronic acid
Molecular Weight (g/mol): 193.01
Molecular Formula: C9H12BNO3
InChi Key: WMMRKCBSJZVRRG-UHFFFAOYSA-N
SMILES: CCNC(=O)C1=CC=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results