accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BOHNCH3CH3H3CClHOOH

CAS RN 850589-46-7

IUPAC Name: [4-(tert-butylcarbamoyl)-3-chlorophenyl]boronic acid
Synonyms: 4-(tert-butylcarbamoyl)-3-chlorophenylboronic acid
Molecular Weight (g/mol): 255.51
Molecular Formula: C11H15BClNO3
InChi Key: RFTCLOPBDGFPDR-UHFFFAOYSA-N
SMILES: CC(C)(C)NC(=O)C1=C(Cl)C=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results