accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BONH2FFFK

CAS RN 850623-63-1

IUPAC Name: potassium (4-carbamoylphenyl)trifluoroboranuide
Synonyms: potassium (4-carbamoylphenyl)trifluoroboranuide
Molecular Weight (g/mol): 227.03
Molecular Formula: C7H6BF3KNO
InChi Key: BMTORUMAAKNFNW-UHFFFAOYSA-N
SMILES: [K+].NC(=O)C1=CC=C(C=C1)[B-](F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results