accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OFF

CAS RN 85068-30-0

IUPAC Name: 1-(2,4-difluorophenyl)propan-1-one
Synonyms: 1-(2,4-difluorophenyl)propan-1-one
Molecular Weight (g/mol): 170.16
Molecular Formula: C9H8F2O
InChi Key: UZWOADNMVRRYDE-UHFFFAOYSA-N
SMILES: CCC(=O)C1=CC=C(F)C=C1F
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
Boiling Point 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (3)
  • (5)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)