accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 856405-77-1

H3CPCH3CH3CH3H3CCH3H3COPH3CCH3H3CCH3H3CCH3CH3

CAS RN 856405-77-1

IUPAC Name: di-tert-butyl[5-(di-tert-butylphosphanyl)-9,9-dimethyl-9H-xanthen-4-yl]phosphane
Synonyms: di-tert-butyl[5-(di-tert-butylphosphanyl)-9,9-dimethylxanthen-4-yl]phosphane
Molecular Weight (g/mol): 498.67
Molecular Formula: C31H48OP2
InChi Key: ZEIZANJFJXHMNS-UHFFFAOYSA-N
SMILES: CC(C)(C)P(C1=CC=CC2=C1OC1=C(C=CC=C1P(C(C)(C)C)C(C)(C)C)C2(C)C)C(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results