missing translation for 'onlineSavingsMsg'
Learn More

4,5-Bis(di-tert-butylphosphino)-9,9-dimethylxanthene, 99%

Product Code. 11327249
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11327249 5 g 5g
11307249 500 mg 500mg
11317249 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11327249 Supplier Thermo Scientific Alfa Aesar Supplier No. H27420.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

CAS: 856405-77-1 | C31H48OP2 | 498.672 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 856405-77-1
Molecular Formula C31H48OP2
Molecular Weight (g/mol) 498.672
MDL Number MFCD07781995
InChI Key ZEIZANJFJXHMNS-UHFFFAOYSA-N
Synonym 9,9-dimethyl-4,5-bis di-tert-butylphosphino xanthene, 9,9-dimethyl-9h-xanthene-4,5-diyl bis di-tert-butylphosphine, t-bu-xantphos, 4,5-bis di-tert-butylphosphino-9,9-dimethylxanthene, 9,9-dimethyl-4,5-bis di-t-butylphosphino xanthene, di-tert-butyl 5-di-tert-butylphosphanyl-9,9-dimethylxanthen-4-yl phosphane, acmc-20alvv, 4,5-bis di-t-butylphosphino-9,9-dimethylxanthene, 4,5-bis di-tert-butylphosphino-9,9-dimethyl-9h-xanthene
PubChem CID 16218202
IUPAC Name ditert-butyl-(5-ditert-butylphosphanyl-9,9-dimethylxanthen-4-yl)phosphane
SMILES CC1(C2=C(C(=CC=C2)P(C(C)(C)C)C(C)(C)C)OC3=C1C=CC=C3P(C(C)(C)C)C(C)(C)C)C

Specifications

Melting Point 153°C to 157°C
Quantity 5 g
Formula Weight 498.68
Percent Purity 99%
Chemical Name or Material 4,5-Bis(di-tert-butylphosphino)-9,9-dimethylxanthene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.