accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONNO

CAS RN 857283-88-6

IUPAC Name: 6-(morpholin-4-yl)pyridine-2-carbaldehyde
Synonyms: 6-(morpholin-4-yl)pyridine-2-carbaldehyde
Molecular Weight (g/mol): 192.22
Molecular Formula: C10H12N2O2
InChi Key: PWJWHXJUKOZOGB-UHFFFAOYSA-N
SMILES: O=CC1=NC(=CC=C1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results