accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3OCH3O

CAS RN 85902-68-7

IUPAC Name: 2-methoxy-5-(propan-2-yl)benzaldehyde
Synonyms: 5-isopropyl-2-methoxybenzaldehyde
Molecular Weight (g/mol): 178.23
Molecular Formula: C11H14O2
InChi Key: FPZHOBMVPBJNKL-UHFFFAOYSA-N
SMILES: COC1=C(C=O)C=C(C=C1)C(C)C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results