missing translation for 'onlineSavingsMsg'
Learn More

5-Isopropyl-2-methoxybenzaldehyde, 97%

Product Code. 11353666
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11353666 100 g 100g
11333666 5 g 5g
11343666 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11353666 Supplier Thermo Scientific Alfa Aesar Supplier No. L14737.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 85902-68-7 | C11H14O2 | 178.231 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85902-68-7
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD03095324
InChI Key FPZHOBMVPBJNKL-UHFFFAOYSA-N
PubChem CID 7009502
IUPAC Name 2-methoxy-5-propan-2-ylbenzaldehyde
SMILES CC(C)C1=CC(=C(C=C1)OC)C=O

Specifications

Boiling Point 96°C (1 mmHg)
Refractive Index 1.539
Quantity 100 g
Beilstein 5923572
Sensitivity Air Sensitive
Synonym 5-isopropyl-2-methoxybenzaldehyde, 2-methoxy-5-isopropylbenzaldehyde, 2-methoxy-5isopropylbenzaldehyde, 2-methoxy-5-methylethyl benzaldehyde, 2-methoxy-5-propan-2-yl-benzaldehyde, 2-methoxy-5-propan-2-yl benzaldehyde, benzaldehyde,2-methoxy-5-1-methylethyl, benzaldehyde, 2-methoxy-5-1-methylethyl
Formula Weight 178.23
Percent Purity 97%
Chemical Name or Material 5-Isopropyl-2-methoxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.