accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 859851-01-7

OSOOH

CAS RN 859851-01-7

IUPAC Name: {2H,3H-thieno[3,4-b][1,4]dioxin-5-yl}methanol
Synonyms: 2H,3H-thieno[3,4-b][1,4]dioxin-5-ylmethanol
Molecular Weight (g/mol): 172.2
Molecular Formula: C7H8O3S
InChi Key: LRWVQOFWMRDMHM-UHFFFAOYSA-N
SMILES: OCC1=C2OCCOC2=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results