accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOOCH3

CAS RN 86-51-1

IUPAC Name: 2,3-dimethoxybenzaldehyde
Synonyms: 2,3-dimethoxybenzaldehyde
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: JIVGSHFYXPRRSZ-UHFFFAOYSA-N
SMILES: COC1=CC=CC(C=O)=C1OC
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)