CAS RN 86309-90-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Chloro-N,N-diethyl-2-nitroaniline, 96%, Thermo Scientific™
CAS: 86309-90-2 Synonym: (4-Chloro-2-nitrophenyl)diethylamine