accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 86404-63-9

FFONNN

CAS RN 86404-63-9

IUPAC Name: 1-(2,4-difluorophenyl)-2-(1H-1,2,4-triazol-1-yl)ethan-1-one
Synonyms: 1-(2,4-difluorophenyl)-2-(1,2,4-triazol-1-yl)ethanone
Molecular Weight (g/mol): 223.18
Molecular Formula: C10H7F2N3O
InChi Key: XCHRPVARHBCFMJ-UHFFFAOYSA-N
SMILES: FC1=CC(F)=C(C=C1)C(=O)CN1C=NC=N1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results