accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

I2H2H2H

CAS RN 865-50-9

IUPAC Name: iodo(²H₃)methane
Synonyms: iodo(²H₃)methane
Molecular Weight (g/mol): 144.96
Molecular Formula: CH3I
InChi Key: INQOMBQAUSQDDS-FIBGUPNXSA-N
SMILES: [2H]C([2H])([2H])I
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results