accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClH3COO(R)NH2SHH

CAS RN 868-59-7

IUPAC Name: hydrogen ethyl (2R)-2-amino-3-sulfanylpropanoate chloride
Synonyms: ethyl cysteinate hcl
Molecular Weight (g/mol): 185.67
Molecular Formula: C5H12ClNO2S
InChi Key: JFKJWWJOCJHMGV-WCCKRBBISA-N
SMILES: [H+].[Cl-].CCOC(=O)[C@@H](N)CS
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (4)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)