Learn More
Thermo Scientific Chemicals L-Cysteine ethyl ester hydrochloride, 98%
Product Code. 10656102 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10656102 25 g 25g
10060850 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10656102 Supplier Thermo Scientific Chemicals Supplier No. 111790250
 Restricted Item
In Stock
Add to basket
CAS: 868-59-7 | C5H12ClNO2S | 185.67 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 868-59-7
Molecular Formula C5H12ClNO2S
MDL Number MFCD00012631
InChI Key JFKJWWJOCJHMGV-WCCKRBBISA-N
Synonym l-cysteine ethyl ester hydrochloride, h-cys-oet.hcl, r-ethyl 2-amino-3-mercaptopropanoate hydrochloride, l-cystein ethyl ester hydrochloride, ethyl l-cysteinate hydrochloride, cysteine ethyl ester hydrochloride, l-cysteine ethyl ester hcl, unii-am467tb2i1, ethyl cysteinate hydrochloride, cysteine, ethyl ester, hydrochloride
PubChem CID 2723617
Specifications
Color White
Loss on Drying 0.5% max. (40°C, 5 hrs) (vacuum, P2O5)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula HSCH2CH(NH2)CO2C2H5·HCl
Quantity 25 g
Specific Rotation -9° to -13° (20°C, 589nm) (c=1, 1M HCl)
SMILES CCOC(=O)C(CS)N.Cl
IUPAC Name ethyl (2R)-2-amino-3-sulfanylpropanoate;hydrochloride
Transmittance 98% min. (c=2.5, H20, 430nm) 1cm cell
Molecular Weight (g/mol) 185.666
Formula Weight 185.67
Percent Purity ≥97.5%
Physical Form Chunks and/or Powder
Chemical Name or Material L-Cysteine ethyl ester hydrochloride, 98%