accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 87-66-1

IUPAC Name: benzene-1,2,3-triol
Synonyms: pyrogallol acid, pyrogallic CI oxidation base 32 pyrogallol (product) fouramine brown AP pyrogallol preparation C.I. oxidation base 32 fourrine PG fourrine 85 pyrogallol (substance)
Molecular Weight (g/mol): 126.11
Molecular Formula: C6H6O3
InChi Key: WQGWDDDVZFFDIG-UHFFFAOYSA-N
SMILES: OC1=CC=CC(O)=C1O
Quantity 
  • (1)
  • (1)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Boiling Point 
  • (8)
  • (6)
Physical Form 
  • (6)
  • (1)
  • (6)
Percent Purity 
  • (2)
  • (3)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (8)
  • (6)
  • (6)
  • (1)
  • (6)
  • (2)
  • (3)
  • (6)
  • (3)
  • (3)