accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BNOCH3H3CHOOH

CAS RN 870521-30-5

IUPAC Name: [6-(propan-2-yloxy)pyridin-3-yl]boronic acid
Synonyms: 6-isopropoxypyridin-3-ylboronic acid
Molecular Weight (g/mol): 181
Molecular Formula: C8H12BNO3
InChi Key: SGEOBUJTURUUJE-UHFFFAOYSA-N
SMILES: CC(C)OC1=NC=C(C=C1)B(O)O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results