accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COHNOOCH3CH3OCH3

CAS RN 87130-20-9

IUPAC Name: propan-2-yl N-(3,4-diethoxyphenyl)carbamate
Synonyms: isopropyl N-(3,4-diethoxyphenyl)carbamate
Molecular Weight (g/mol): 267.33
Molecular Formula: C14H21NO4
InChi Key: LNJNFVJKDJYTEU-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(NC(=O)OC(C)C)C=C1OCC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results