missing translation for 'onlineSavingsMsg'
Learn More

Diethofenacarb, TRC

Product Code. 30012148
Change view
Click to view available options
Quantity:
250 mg
1 g
25 g
Unit Size:
1g
250mg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012148 25 g 25g
17954765 250 mg 250mg
17970575 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012148 Supplier Toronto Research Chemicals Supplier No. TRCD44190525G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87130-20-9
Molecular Formula C14 H21 N O4
Molecular Weight (g/mol) 267.32
Synonym Carbamic acid, (3,4-diethoxyphenyl)-, 1-methylethyl ester (9CI), Diethofencarb, Isopropyl 3,4-diethoxycarbanilate, Isopropyl N-(3,4-diethoxyphenyl)carbamate, Powmil, Powmyl, S 32165
IUPAC Name propan-2-yl N-(3,4-diethoxyphenyl)carbamate
SMILES CCOc1ccc(NC(=O)OC(C)C)cc1OCC

Specifications

Quantity 25 g
InChI Formula InChI=1S/C14H21NO4/c1-5-17-12-8-7-11(9-13(12)18-6-2)15-14(16)19-10(3)4/h7-10H,5-6H2,1-4H3,(H,15,16)
Formula Weight 267.1471
Percent Purity >95%
Chemical Name or Material Diethofencarb
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.