CAS RN 876107-33-4
IUPAC Name:
λ⁴-cerium(4+) tetrakis(2-methoxyethan-1-olate)
Synonyms:
λ⁴-cerium(4+) tetrakis(2-methoxyethanolate)
Molecular Weight (g/mol):
440.46
Molecular Formula:
C12H28CeO8
InChi Key:
JRRALIVAHBLZSS-UHFFFAOYSA-N
SMILES:
[Ce+4].COCC[O-].COCC[O-].COCC[O-].COCC[O-]
1
–
1
of
1
results
1
Cerium(IV) 2-methoxyethoxide, 18-20% w/w in 2-methoxyethanol
CAS: 876107-33-4 Molecular Formula: C12H28CeO8 Molecular Weight (g/mol): 440.464 MDL Number: MFCD00236530 InChI Key: JRRALIVAHBLZSS-UHFFFAOYSA-N Synonym: cerium iv methoxyethoxide w/w in methoxyethanol,tetrakis 2-methoxyethoxy cerium iv,cerium 4+ tetrakis 2-methoxyethan-1-olate,cerium iv methoxyethoxide in methoxyethanol PubChem CID: 21200429 IUPAC Name: cerium(4+);2-methoxyethanolate SMILES: COCC[O-].COCC[O-].COCC[O-].COCC[O-].[Ce+4]