CAS RN 877-88-3
IUPAC Name:
1-(bromomethyl)-3,5-dimethoxybenzene
Synonyms:
1-(bromomethyl)-3,5-dimethoxybenzene
Molecular Weight (g/mol):
231.09
Molecular Formula:
C9H11BrO2
InChi Key:
BTHIGJGJAPYFSJ-UHFFFAOYSA-N
SMILES:
COC1=CC(OC)=CC(CBr)=C1
1
–
2
of
2
results
1
Promotions Available
3,5-Dimethoxybenzyl bromide, 95%
3,5-Dimethoxybenzyl bromide, 95%
2
3,5-Dimethoxybenzyl Bromide, TRC
CAS: 877-88-3 Molecular Formula: C9H11BrO2 Molecular Weight (g/mol): 231.09 Synonym: 1-(Bromomethyl)-3,5-dimethoxy-benzene,5-(Bromomethyl)-1,3-dimethoxybenzene,alpha-Bromo-3,5-dimethoxytoluene SMILES: COc1cc(CBr)cc(OC)c1