missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxybenzyl Bromide, TRC

Product Code. 30386858
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30386858 50 g
30376703 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30386858 Supplier Toronto Research Chemicals Supplier No. TRCD46083550G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 877-88-3
Molecular Formula C9H11BrO2
Molecular Weight (g/mol) 231.09
Synonym 1-(Bromomethyl)-3,5-dimethoxy-benzene, 5-(Bromomethyl)-1,3-dimethoxybenzene, alpha-Bromo-3,5-dimethoxytoluene
SMILES COc1cc(CBr)cc(OC)c1

Specifications

Quantity 50 g
InChI Formula InChI=1S/C9H11BrO2/c1-11-8-3-7(6-10)4-9(5-8)12-2/h3-5H,6H2,1-2H3
Formula Weight 229.9942
Chemical Name or Material 3,5-Dimethoxybenzyl Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.