CAS RN 88426-33-9
CAS RN 88426-33-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Buparvaquone, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Buparvaquone, TRC
CAS: 88426-33-9 Molecular Formula: C21 H26 O3 Molecular Weight (g/mol): 326.43 Synonym: 2-[[4-(1,1-Dimethylethyl)cyclohexyl]methyl]-3-hydroxy-1,4-naphthalenedione,Buparvaquone,Butalex IUPAC Name: 2-[(4-tert-butylcyclohexyl)methyl]-3-hydroxynaphthalene-1,4-dione SMILES: CC(C)(C)C1CCC(CC2=C(O)C(=O)c3ccccc3C2=O)CC1