missing translation for 'onlineSavingsMsg'
Learn More

Buparvaquone, TRC

Product Code. 17992345
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17992345 5 mg 5mg
17961475 25 mg 25mg
17922015 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17992345 Supplier Toronto Research Chemicals Supplier No. TRCB6895555MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88426-33-9
Molecular Formula C21 H26 O3
Molecular Weight (g/mol) 326.43
Synonym 2-[[4-(1,1-Dimethylethyl)cyclohexyl]methyl]-3-hydroxy-1,4-naphthalenedione, Buparvaquone, Butalex
IUPAC Name 2-[(4-tert-butylcyclohexyl)methyl]-3-hydroxynaphthalene-1,4-dione
SMILES CC(C)(C)C1CCC(CC2=C(O)C(=O)c3ccccc3C2=O)CC1

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C21H26O3/c1-21(2,3)14-10-8-13(9-11-14)12-17-18(22)15-6-4-5-7-16(15)19(23)20(17)24/h4-7,13-14,24H,8-12H2,1-3H3
Formula Weight 326.1882
Chemical Name or Material Buparvaquone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.