accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHO

CAS RN 90-02-8

IUPAC Name: 2-hydroxybenzaldehyde
Synonyms: salicylaldehyde salicylaldehyd O-hydroxybenzaldehyde salicylaldehyde (product) salicylal salizylaldehyd salicyladehyde salicylaldehyde (substance)
Molecular Weight (g/mol): 122.12
Molecular Formula: C7H6O2
InChi Key: SMQUZDBALVYZAC-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1C=O
Percent Purity 
  • (3)
  • (7)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results