accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 90-99-3

IUPAC Name: [chloro(phenyl)methyl]benzene
Synonyms: [chloro(phenyl)methyl]benzene
Molecular Weight (g/mol): 202.68
Molecular Formula: C13H11Cl
InChi Key: ZDVDCDLBOLSVGM-UHFFFAOYSA-N
SMILES: ClC(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (6)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)