CAS RN 9001-75-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
2
MP Biomedicals™ Pepsin
Pepsin is a peptidase used to digest proteins and is commonly used in the preparation of Fab fragments from antibodies. Pepsin, from porcine gastric mucosa, has been used to hydrolyze dry cervical samples in mice
3
Thermo Scientific™ Pepsin from Porcine Stomach
Derived from Porcine stomach