missing translation for 'onlineSavingsMsg'
Learn More

Pepsin Powder, Purified Laboratory Grade, J.T. Baker™

Product Code. 15598054 Shop All J.T.Baker Products
Change view
Click to view available options
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15598054 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15598054 Supplier JT Baker Supplier No. 284401

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For Laboratory, Research, or Manufacturing Use

Specifications

CAS 9001-75-6
Assay Percent Range 2000 - 3500 pepsin units/mg
Packaging Poly Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.