accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

∗FF∗FFn

CAS RN 9002-84-0

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: (C2F4)n
InChi Key: —
SMILES: FC(F)(-*)C(F)(F)-*
Molecular Weight (g/mol) 
  • (3)
  • (17)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (5)
  • (3)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (14)
  • (3)
  • (17)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (5)
  • (3)
  • (1)
  • (2)