missing translation for 'onlineSavingsMsg'
Learn More

Polytetrafluoroethylene

Product Code. 11460433
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460433 250 g 250g
11450433 50 g 50g
11444354 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460433 Supplier Thermo Scientific Alfa Aesar Supplier No. A12613.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 9002-84-0 | (C2F4)n | NaN g/mol

PTFE may be used as a non stick coating. It can be used as engineering plastics. It is employed as lining reaction vessels, gaskets and pump packing, electrical insulator for high frequency applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-84-0
Molecular Formula (C2F4)n
Molecular Weight (g/mol) NaN
MDL Number MFCD00084455
SMILES FC(F)(-*)C(F)(F)-*

Specifications

Merck Index 14,7562
Solubility Information Insoluble in water.
Quantity 250 g
Chemical Name or Material Polytetrafluoroethylene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.