accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

∗∗OHn

CAS RN 9002-89-5

IUPAC Name: ethenol
Synonyms: vinyl alcohol mowiol
Molecular Weight (g/mol): 44.05
Molecular Formula: C2H4O
InChi Key: IMROMDMJAWUWLK-UHFFFAOYSA-N
SMILES: OC=C
Quantity 
  • (1)
  • (14)
  • (1)
  • (16)
  • (8)
  • (13)
  • (2)
  • (1)
  • (1)
  • (10)
Percent Purity 
  • (6)
  • (3)
  • (1)
  • (4)
  • (5)
  • (3)
  • (1)
  • (3)
  • (9)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (4)
  • (40)
  • (18)
  • (3)
  • (16)
  • (1)
  • (14)
  • (1)
  • (16)
  • (8)
  • (13)
  • (2)
  • (1)
  • (1)
  • (10)
  • (6)
  • (3)
  • (1)
  • (4)
  • (5)
  • (3)
  • (1)
  • (3)
  • (9)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)