CAS RN 9002-93-1
Triton X-100
CAS: 9002-93-1 Molecular Formula: C16H26O2 Molecular Weight (g/mol): 250.38 MDL Number: MFCD00132505 InChI Key: JYCQQPHGFMYQCF-UHFFFAOYSA-N PubChem CID: 5590 SMILES: CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1
Triton™ X-100 (Electrophoresis), Fisher BioReagents™
CAS: 9002-93-1 Molecular Formula: C16H26O2 Molecular Weight (g/mol): 250.38 MDL Number: MFCD00132505 InChI Key: JYCQQPHGFMYQCF-UHFFFAOYSA-N Synonym: Polyethylene Glycol p-tert-Octylphenyl Ether PubChem CID: 5590 SMILES: CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1
IGEPAL∣r CA-630
CAS: 9002-93-1 Molecular Formula: C16H26O2 Molecular Weight (g/mol): 250.38 MDL Number: MFCD00132505 InChI Key: JYCQQPHGFMYQCF-UHFFFAOYSA-N Synonym: Octylphenyl-polyethylene glycol IUPAC Name: 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethan-1-ol SMILES: CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1
Thermo Scientific Chemicals Triton™ X-114
CAS: 9002-93-1 Molecular Formula: C16H26O2 Molecular Weight (g/mol): 250.38 MDL Number: MFCD00132505 InChI Key: JYCQQPHGFMYQCF-UHFFFAOYSA-N Synonym: Polyoxyethylene(8) octylphenyl ether PubChem CID: 5590 IUPAC Name: 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethanol SMILES: CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1
Thermo Scientific Chemicals Triton∣r X-100, Electrophoresis Reagent
CAS: 9002-93-1 Molecular Formula: C16H26O2 Synonym: Octoxynol-9; Polyethylene glycol tert-octylphenyl ether
Nonidet™ P40 Substitute, Ultrapure, Thermo Scientific Chemicals
CAS: 9002-93-1 Molecular Formula: C16H26O2 Molecular Weight (g/mol): 250.38 MDL Number: MFCD00132505 InChI Key: JYCQQPHGFMYQCF-UHFFFAOYSA-N Synonym: Igepal CA to 630,Polyethylene glycol tert to octylphenyl ether PubChem CID: 5590 IUPAC Name: 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethanol SMILES: CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1
Melford Nonidet P40 Substitute
Laboratory reagent grade.
R&D Systems™ 10X Strep-Diluent
R&D Systems™ 10X Strep-Diluent is a solution that is used as a diluent for the Strep-HRP.
Triton X-100, TRC
CAS: 9002-93-1 Molecular Formula: C14H22O.(OCH2CH2)n Molecular Weight (g/mol): 250.38 Synonym: Polyethyleneglycols Mono[p-(1,1,3,3-tetramethylbutyl)phenyl] Ether,p-(1,1,3,3-Tetramethylbutyl)phenol Monoether with Polyethylene Glycol,(p-t-Octylphenoxy)polyethoxyethanol,Anapoe X 100,Anapoe X 114,Antarox A 200,Coulter Dispersant IA,Criton X 100,Hydrol SW,Iconol OP,Intra-Cell,Koromex II,Lutensol OP 10,NOP 90,Nonidel P 40,OPE 30,Octoxynol 5,Ortho-Gynol,Photo-Flow 200,Poly(oxyethylene) p-tert-Octylphenyl Ether,Polyethylene Gllycol Mono(4-octylphenyl) Ether,Polyethylene Glycol Mono(4-tert-octylphenyl) Ether,Polyoxyethylene Glycol-p-tert-octylphenyl Ether,Polyoxyethylene Mono(octylphenyl) Ether,Preceptin,Surfac OP 30,Surfac OP 5,Surfonic OP 120,TOP 10,TX 100,TX 102,TX 305,TX 35,TX 405,Texofor FP 300,Octoxynol 9,Octylphenoxy poly(ethyleneoxy)ethanol, branched IUPAC Name: 14-(4-(2,4,4-trimethylpentan-2-yl)phenoxy)-3,6,9,12-tetraoxatetradecan-1-ol SMILES: CC(C(C=C1)=CC=C1OCCOCCOCCOCCOCCO)(C)CC(C)(C)C
Triton™ X-100, Extra Pure, SLR, Fisher Chemical™
For solubilization and permeabilization of membranes
Thermo Scientific Chemicals Triton™ X-405, 70% solution in water
CAS: 9002-93-1 | C16H26O2 | 250.38 g/mol
Triton™ X-100, Thermo Scientific™
CAS: 9002-93-1 Molecular Formula: C16H26O2 Molecular Weight (g/mol): 250.38 MDL Number: MFCD00132505 InChI Key: JYCQQPHGFMYQCF-UHFFFAOYSA-N Synonym: Polyoxyethylene(10) octylphenyl ether PubChem CID: 5590 SMILES: CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1
Thermo Scientific Chemicals Triton™ X-100, 98%, for molecular biology, DNAse, RNAse and Protease free
CAS: 9002-93-1 Molecular Formula: C16H26O2 Synonym: Polyethylene glycol mono [4-(1,1,3,3-tetramethylbutyl)phenyl] ether
- 1
- 2