missing translation for 'onlineSavingsMsg'
Learn More

Triton™ X-100, Extra Pure, SLR, Fisher Chemical™

Product Code. 10509020 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Unit Size:
2.5 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10509020 2.5 L 2.5 L
10254640 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 10509020 Supplier Fisher Chemical Supplier No. T/3751/17
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

For solubilization and permeabilization of membranes

This nonionic detergent is used to solubilize membranes, permeabilize unfixed eukaryotic membranes, and is a component of lysis buffers for DNA extraction.
TRUSTED_SUSTAINABILITY

Especificaciones

Color Clear Yellow
CAS 9002-93-1
Molecular Formula C34H62O11
Formula Weight 646.86g/mol
Quantity 2.5 L
compliance-icons compliance-icons
Nombre del producto
  • Triton X-100
Palabra de advertencia
  • Danger
Clasificación
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Indicaciones de peligro
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.