accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 9003-11-6

CH3OO

CAS RN 9003-11-6

IUPAC Name: 2-methyloxirane; oxirane
Synonyms: poloxamer 101 meroxapol 105 meroxapol 108 meroxapol 171 meroxapol 172 meroxapol 174 meroxapol 178 meroxapol 251 meroxapol 252 meroxapol 254
Molecular Weight (g/mol): 102.13
Molecular Formula: C5H10O2
InChi Key: RVGRUAULSDPKGF-UHFFFAOYNA-N
SMILES: C1CO1.CC1CO1
Quantity 
  • (1)
  • (1)
  • (1)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results