accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNOOH3CNNNNOONH3C(E/Z)

CAS RN 9004-57-3

IUPAC Name: 2-[butyl({4-[2-(2,6-dicyano-4-nitrophenyl)diazen-1-yl]-3-methylphenyl})amino]ethyl acetate
Synonyms: ethylcellulose ethyl cellulose
Molecular Weight (g/mol): 448.48
Molecular Formula: C23H24N6O4
InChi Key: ARSKJXYLLONUAJ-UHFFFAOYSA-N
SMILES: CCCCN(CCOC(C)=O)C1=CC(C)=C(C=C1)N=NC1=C(C=C(C=C1C#N)[N+]([O-])=O)C#N
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (5)