missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ethyl cellulose

Product Code. 15413737
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15413737 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15413737 Supplier Thermo Scientific Chemicals Supplier No. H27621.MD
  • ONLINE EXCLUSIVE PRICE
    102.00€
    91.65€ / 250mg
    Save 10.35€
    10% Off
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 9004-57-3 | C23H24N6O4 | 448.483 g/mol

Ethyl cellulose is used as a food additive and as an emulsifier. It is also used as a film-former in coatings, hot-melt adhesives and transfer inks. Further, it is employed as a binder, filler, plastic coating and diluents of color. In addition to this, it is used in lacquers and plastic toughing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl cellulose is used as a food additive and as an emulsifier. It is also used as a film-former in coatings, hot-melt adhesives and transfer inks. Further, it is employed as a binder, filler, plastic coating and diluents of color.
TRUSTED_SUSTAINABILITY

Specifications

CAS 9004-57-3
Density 1.14
Molecular Formula C23H24N6O4
Refractive Index 1.47
MDL Number MFCD00131037
Quantity 250 mg
Synonym n-butyl-n-2-acetoxyethyl-4-4-nitro-2,6-dicyanophenyl azo-3-methylbenzeneamine, 1,3-benzenedicarbonitrile, 2-4-2-acetyloxy ethyl butylamino-2-methylphenyl azo-5-nitro, 1,3-benzenedicarbonitrile, 2-2-4-2-acetyloxy ethyl butylamino-2-methylphenyl diazenyl-5-nitro, azo dye n5, 4-2,6-dicyano-4-nitrophenyl azo-3-methylphenyl amino hexyl acetate, ethyl cellulose, viscosity ethoxyl, 2-n-butyl-4-2,6-dicyano-4-nitrophenyl diazenyl-3-methylanilino ethyl acetate, 4-n-2-acetoxyethyl butyl amino-2',6'-dicyano-2-methyl-4'-nitroazobenzene
Solubility Information Soluble in tetrahydrofuran,methyl acetate,ethanol,methanol,toluene,ethyl acetate,chloroform,esters,aromatic hydrocarbons,alcohols and ketones. Insoluble in water,glycerol and propane-1,2-diol.
InChI Key ARSKJXYLLONUAJ-UHFFFAOYSA-N
SMILES CCCCN(CCOC(=O)C)C1=CC(=C(C=C1)N=NC2=C(C=C(C=C2C#N)[N+](=O)[O-])C#N)C
IUPAC Name 2-[N-butyl-4-[(2,6-dicyano-4-nitrophenyl)diazenyl]-3-methylanilino]ethyl acetate
Molecular Weight (g/mol) 448.483
PubChem CID 166304
Chemical Name or Material Ethyl cellulose
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.