accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 9013-20-1

NClSOOHBr

CAS RN 9013-20-1

IUPAC Name: 1-{[(carboxymethyl)sulfanyl](3-chlorophenyl)methylidene}-1λ⁵-piperidin-1-ylium bromide
Synonyms: 1-{[(carboxymethyl)sulfanyl](3-chlorophenyl)methylidene}-1λ⁵-piperidin-1-ylium bromide
Molecular Weight (g/mol): 378.71
Molecular Formula: C14H17BrClNO2S
InChi Key: RTWACOLFHOBGCE-UHFFFAOYSA-N
SMILES: [Br-].OC(=O)CSC(C1=CC(Cl)=CC=C1)=[N+]1CCCCC1
Color 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results