accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3HNOOH3C

CAS RN 902836-89-9

IUPAC Name: 1-(2-methoxyphenyl)-2-[(propan-2-yl)amino]ethan-1-one
Synonyms: 2-(isopropylamino)-1-(2-methoxyphenyl)ethanone
Molecular Weight (g/mol): 207.27
Molecular Formula: C12H17NO2
InChi Key: FGSJNHHRBVOFTM-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1C(=O)CNC(C)C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results