accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HClNH2FFF

CAS RN 90774-69-9

IUPAC Name: hydrogen 4-(trifluoromethyl)aniline chloride
Synonyms: hydrogen P-trifluoromethylaniline chloride
Molecular Weight (g/mol): 197.59
Molecular Formula: C7H7ClF3N
InChi Key: CZGDYZDUMJDQSD-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC1=CC=C(C=C1)C(F)(F)F
Molecular Weight (g/mol) 
  • (6)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results