missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzotrifluoride hydrochloride, 97%, ACROS Organics™

Product Code. 10377350 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10377350 5 g Glass bottle 5g
10194090 1 g Glass bottle 1g
10295840 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10377350 Supplier Thermo Scientific Acros Supplier No. 174180050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 90774-69-9
Molecular Formula C7H7ClF3N
Molecular Weight (g/mol) 197.585
MDL Number MFCD00007897
InChI Key CZGDYZDUMJDQSD-UHFFFAOYSA-N
Synonym 4-aminobenzotrifluoride hydrochloride, 4-trifluoromethyl aniline hydrochloride, 4-trifluoromethyl anilinium chloride, p-trifluoromethylaniline hydrochloride, 4-trifluoromethyl phenylamine, chloride, pubchem2816, 4-aminobenzotrifluoride, hydrochloride, 4-trifluoromethylaniline hydrochloride, 4-trifluoromethyl aniline;hydrochloride, a,a,a-trifluoro-p-toluidine hydrochloride
PubChem CID 2723961
IUPAC Name 4-(trifluoromethyl)aniline;hydrochloride
SMILES C1=CC(=CC=C1C(F)(F)F)N.Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula H2NC6H4CF3·HCl
Quantity 5 g
Formula Weight 197.59
Percent Purity ≥96.0%
Physical Form Granular Powder
Chemical Name or Material 4-Aminobenzotrifluoride hydrochloride, 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.