accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BOH3COHHOOH

CAS RN 908142-03-0

IUPAC Name: [3-(hydroxymethyl)-4-methoxyphenyl]boronic acid
Synonyms: 3-(hydroxymethyl)-4-methoxyphenylboronic acid
Molecular Weight (g/mol): 181.98
Molecular Formula: C8H11BO4
InChi Key: DOQUCBSZQOPLGT-UHFFFAOYSA-N
SMILES: COC1=C(CO)C=C(C=C1)B(O)O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results