accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 90982-32-4

CH3OOSOOHNOHNNClOH3CN

CAS RN 90982-32-4

IUPAC Name: ethyl 2-({[(4-chloro-6-methoxypyrimidin-2-yl)carbamoyl]amino}sulfonyl)benzoate
Synonyms: chlorimuron-ethyl
Molecular Weight (g/mol): 414.82
Molecular Formula: C15H15ClN4O6S
InChi Key: NSWAMPCUPHPTTC-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=CC=C1S(=O)(=O)NC(=O)NC1=NC(Cl)=CC(OC)=N1
Color 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)