Learn More
Chlorimuron ethyl, 95%, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar J66605.06
| Quantity | 5g |
|---|
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 90982-32-4 | |
| 414.817 | |
| NSWAMPCUPHPTTC-UHFFFAOYSA-N | |
| 56160 | |
| ethyl 2-[(4-chloro-6-methoxypyrimidin-2-yl)carbamoylsulfamoyl]benzoate |
| C15H15ClN4O6S | |
| MFCD00128063 | |
| Ethyl 2-(4-chloro-6-methoxy-2-pyrimidinylcarbamoylsulfamoyl)benzoate | |
| CHEBI:47319 | |
| CCOC(=O)C1=CC=CC=C1S(=O)(=O)NC(=O)NC2=NC(=CC(=N2)Cl)OC |
Specifications
| 90982-32-4 | |
| 5g | |
| MFCD00128063 | |
| Ethyl 2-(4-chloro-6-methoxy-2-pyrimidinylcarbamoylsulfamoyl)benzoate | |
| NSWAMPCUPHPTTC-UHFFFAOYSA-N | |
| ethyl 2-[(4-chloro-6-methoxypyrimidin-2-yl)carbamoylsulfamoyl]benzoate | |
| 56160 | |
| 414.82 | |
| Chlorimuron ethyl |
| Colorless | |
| C15H15ClN4O6S | |
| 14,2093 | |
| Freely soluble in aqueous solution | |
| CCOC(=O)C1=CC=CC=C1S(=O)(=O)NC(=O)NC2=NC(=CC(=N2)Cl)OC | |
| 414.817 | |
| CHEBI:47319 | |
| 95% |
Safety and Handling
- Chlorimuron ethyl
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
Hazard Statement
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P391-Collect spillage.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contain: Chlorimuron ethyl
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.