accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2N

CAS RN 91-00-9

IUPAC Name: 1,1-diphenylmethanamine
Synonyms: benzhydrylamin
Molecular Weight (g/mol): 183.25
Molecular Formula: C13H13N
InChi Key: MGHPNCMVUAKAIE-UHFFFAOYSA-N
SMILES: NC(C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results