accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 91-58-7

IUPAC Name: 2-chloronaphthalene
Synonyms: 2-chloronaphthalene
Molecular Weight (g/mol): 162.62
Molecular Formula: C10H7Cl
InChi Key: CGYGETOMCSJHJU-UHFFFAOYSA-N
SMILES: ClC1=CC=C2C=CC=CC2=C1
Molecular Weight (g/mol) 
  • (4)
  • (5)
Quantity 
  • (1)
  • (2)
  • (2)
Solvent or Matrix 
  • (1)
  • (1)
  • (1)
  • (1)
Concentration 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results