missing translation for 'onlineSavingsMsg'
Learn More

Metabolite 2-Chloronaphthalene in Acetonitrile 10μg/mL, Fisher Chemical™

Product Code. 12942464 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12942464 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12942464 Supplier Fisher Chemical Supplier No. P811590

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Packaging Ampoule(s)
CAS 91-58-7
Molecular Formula C10H7Cl
Concentration 10 μg/mL
UN Number 1648
InChI Key CGYGETOMCSJHJU-UHFFFAOYSA-N
SMILES C1=CC=C2C=C(C=CC2=C1)Cl
IUPAC Name 2-chloronaphthalene
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 162.616
PubChem CID 7056
Quantity 10 mL
Chemical Name or Material 2-Chloronaphthalene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Standard solution 2-Chloronaphthalene 10ug/ml in acetonitrile
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.