accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 91-95-2

IUPAC Name: [1,1'-biphenyl]-3,3',4,4'-tetramine
Synonyms: 3,3'-diaminobenzidine diaminobenzidine
Molecular Weight (g/mol): 214.27
Molecular Formula: C12H14N4
InChi Key: HSTOKWSFWGCZMH-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1N)C1=CC=C(N)C(N)=C1
Molecular Weight (g/mol) 
  • (5)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (5)
  • (5)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)